Test charts
Test charts, hvid/sort, 12G
Catalog # SIMEVF3007
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
- Dimensions from A6 up to A4
- Film laminated for an excellent solvent, chemical resistance, and film spread
- Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Ordering information: A certificate is available for charts with an optical brightener.
Documents
Specifications
- Format:12G
- Farve:hvid/sort
- Beskrivelse:Test charts
- Seq:18