Accessories for dosing pump, Simdos®, KNF
In-line filter FS 25 T PVDF mesh opening 70 µm SIMDOS® 10
Katalog # KNFA165211
Leverandør: KNF
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:In-line filter FS 25 T PVDF mesh opening 70 µm SIMDOS® 10
- Seq:03