Orders LinkContactUsLinkComponent
Temperature control systems, PRESTO®
Temperature control systems, PRESTO®
W91x, –91...+250 °C, ±0,05...±0,2 °C, 18 kW, 11,0; 11,0; 11,0; 10,0; 9,5; 9,0; 6,5; 1,5 kW, R404A/R449A, 950×1270×1900 mm, 870 kg
Katalog # 461-0807
Leverandør:   JULABO GmbH
Temperature control systems, PRESTO®
Katalog # 461-0807
Leverandør:  JULABO GmbH
Temperature control systems, PRESTO®
Temperature control systems, PRESTO®
W91x, –91...+250 °C, ±0,05...±0,2 °C, 18 kW, 11,0; 11,0; 11,0; 10,0; 9,5; 9,0; 6,5; 1,5 kW, R404A/R449A, 950×1270×1900 mm, 870 kg
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exothermic and endothermic reactions with extraordinary speed, they feature high flow rates, constant pressure and a controlled build-up of pump pressure. Changes in the temperature control of liquid viscosity are balanced dynamically. Permanent internal monitoring and self-lubricating pumps contribute to long service life. The integrated touch panel gives the user a clear view of important information. Fingertip control via the touch screen. Multiple interfaces enable remote control of the PRESTO® across networks and in super-ordinated control systems. Wireless remote control of the new PRESTO® can easily be done by using the Wireless TEMP range of products.
  • Fingertip control via the touch screen with 14,4 cm colour touch panel
  • Up to three user levels with password management
  • USB (Host und Device), Ethernet and SD card slot
  • Units are whisper-quiet and barely audible when operating
  • Filling system is accessible from the top
  • Choice of models with environmentally-friendly natural refrigerant
Max. flow rate/max. pressure:
A30: 25 L/min, 0,5 bar
A38/A41/W41: 0 to 50 L/min, 0,1 to 1,6 bar
A40/W40, A80/W80: 16 to 40 L/min, 0,3 to 1,7 bar
A45/A45t/W50/W50t: 35 to 76 L/min, 0,48 to 3,2 bar
A70: 0 to 40 L/min, 0,1 to 1,7 bar W91/W92 (t/tt): 26 to 80 L/min, 0,5 to 3,0 bar W91×/W92× (t/tt): 18 to 70 L/min, 0,8 to 5,5 bar
W93: 0 to 80 L/min, 0,1 to 3 bar W93x: 0 to 70 L/min, 0,1 to 5,5 bar.
Ordering information: Supplied without accessories. Go to website for details of accessories.
Caution: Cooling capacity at +200, +100, +20, 0, –10, –20, –30 and −40 °C. W91/W92/W93 (t/tt/x): Cooling capacity at +200, +100, +20, 0, –20, –40, –60 and −80 °C.
Specifikationer
  • Opvarmnings kapacitet:
    18 kW
  • Temperaturområde:
    –91...+250 °C
  • Vægt:
    870 kg
  • Seq:
    025
  • B×D×H:
    950×1270×1900 mm
  • Refrigerant:
    R404A/R449A
  • Kølekapacitet:
    11,0; 11,0; 11,0; 10,0; 9,5; 9,0; 6,5; 1,5 kW
  • Temp. stabilitet:
    ±0,05...±0,2 °C
  • Model:
    W91x
Accessories
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to M16×1 male
    2× adapters, M24×1,5 female to M16×1 male
    JULA8890052
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.210,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to tube ½"
    2× adapters, M24×1,5 female to tube ½"
    JULA8890065
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.600,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to tube 1"
    2× adapters, M24×1,5 female to tube 1"
    JULA8890066
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.600,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to M24×1,5 female
    2× adapters, M24×1,5 female to M24×1,5 female
    JULA8890067
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.760,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to M30×1,5 male
    2× adapters, M24×1,5 female to M30×1,5 male
    JULA8890068
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.940,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 male to M30×1,5 female
    2× adapters, M24×1,5 male to M30×1,5 female
    JULA8890069
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 2.160,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to M30×1,5 female
    2× adapters, M24×1,5 female to M30×1,5 female
    JULA8890070
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 2.330,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 male to M16×1 female
    2× adapters, M24×1,5 male to M16×1 female
    JULA8890071
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.980,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to NPT ¾" female
    2× adapters, M30×1,5 female to NPT ¾" female
    JULA8890084
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.700,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Profibus DP interface, for instruments with RS232
    Profibus DP interface, for instruments with RS232
    JULA8900020
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 11.530,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • RS485 interface, for instruments with RS232
    RS485 interface, for instruments with RS232
    JULA8900024
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 11.530,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Pressure sensor, 2 fittings M30×1,5 male (−95 ... +250 °C)
    Pressure sensor, 2 fittings M30×1,5 male (−95 ... +250 °C)
    JULA8980772
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 4.540,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Pressure sensor, 2 fittings M38×1,5 male (−95 ... +250 °C)
    Pressure sensor, 2 fittings M38×1,5 male (−95 ... +250 °C)
    JULA8980773
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 5.180,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Electronic module with analog connectors (input, output, standby-in)
    Electronic module with analog connectors (input, output, standby-in)
    JULA8900105
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 4.330,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Pressure sensor, 2 fittings M24×1,5 male (−95 ... +250 °C)
    Pressure sensor, 2 fittings M24×1,5 male (−95 ... +250 °C)
    JULA8980771
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 4.370,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to tube ¾"
    2× adapters, M24×1,5 female to tube ¾"
    JULA8890064
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.110,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to M38×1,5 male
    2× adapters, M30×1,5 female to M38×1,5 male
    JULA8890080
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.970,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 male to M38×1,5 female
    2× adapters, M30×1,5 male to M38×1,5 female
    JULA8890081
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.970,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to NPT ¾" male
    2× adapters, M30×1,5 female to NPT ¾" male
    JULA8890083
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.920,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to NPT 1" male
    2× adapters, M30×1,5 female to NPT 1" male
    JULA8890085
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.700,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to NPT 1" female
    2× adapters, M30×1,5 female to NPT 1" female
    JULA8890086
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.860,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to tube 1"
    2× adapters, M30×1,5 female to tube 1"
    JULA8890087
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.500,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to tube ¼"
    2× adapters, M24×1,5 female to tube ¼"
    JULA8890063
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.110,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to M38×1,5 female
    2× adapters, M30×1,5 female to M38×1,5 female
    JULA8890082
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 2.210,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Sight glass, −100…+280 °C, PN16/Class 230, 2 fittings M30×1,5 male
    Sight glass, −100…+280 °C, PN16/Class 230, 2 fittings M30×1,5 male
    JULA8970815
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 11.410,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT 1" male
    2× adapters, M24×1,5 female to NPT 1" male
    JULA8890062
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.690,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Adapter, M24×1,5 male to M24×1,5 male
    Adapter, M24×1,5 male to M24×1,5 male
    JULA8890110
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 646,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ¾" male
    2× adapters, M24×1,5 female to NPT ¾" male
    JULA8890061
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.690,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ½" male
    2× adapters, M24×1,5 female to NPT ½" male
    JULA8890060
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.690,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ³/₈" male
    2× adapters, M24×1,5 female to NPT ³/₈" male
    JULA8890059
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.150,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ¼" male
    2× adapters, M24×1,5 female to NPT ¼" male
    JULA8890058
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.080,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT 1" female
    2× adapters, M24×1,5 female to NPT 1" female
    JULA8890057
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 2.160,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ¾" female
    2× adapters, M24×1,5 female to NPT ¾" female
    JULA8890056
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.920,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ½" female
    2× adapters, M24×1,5 female to NPT ½" female
    JULA8890055
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.710,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ³/₈" female
    2× adapters, M24×1,5 female to NPT ³/₈" female
    JULA8890054
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.710,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M24×1,5 female to NPT ¼" female
    2× adapters, M24×1,5 female to NPT ¼" female
    JULA8890053
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 1.210,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to M38×1,5 female
    2× adapters, M30×1,5 female to M38×1,5 female
    JULA8890089
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 3.240,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • 2× adapters, M30×1,5 female to M30×1,5 female
    2× adapters, M30×1,5 female to M30×1,5 female
    JULA8890088
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 2 Items - 2.380,00 kr
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • EtherCAT interface is a protocol converter from EtherCAT to RS232
    EtherCAT interface is a protocol converter from EtherCAT to RS232
    JULA8900025
    All New Presto® series units
    Log ind for tilgængelighed
    1 * 1 Items - 11.530,00 kr