Venepunktursæt, Surshield™ Surflo®
Winged needles with protector, G 21, grøn, 19 mm
Catalog # TERUSV-S21NL30
Supplier: TERUMO
Jump to:
Product Details & Documents
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.
Specifications
- Måler:G 21
- Farve:grøn
- Beskrivelse:Winged needles with protector
- Længde:19 mm
- Seq:002