Cleaning system for controlled environment
CE bucket system trolley, without accessories, suitable for autoclaving and ISO Class 5
141-0937
These cleanroom cleaning trolleys are specially designed for use in sensitive areas and controlled environments. The cleaning trolleys are extremely functional, versatile and are characterised by superior ergonomic properties.
- Small and compact
- Heat and chemical resistant
- With electrically conductive castors, the tread of the wheels is made from an hydrolysis resistant material
- Fully autoclavable and suitable for gamma sterilisation
ISO 5 / GMP A + B / D FED209 class 100
Certifications: ISO 5 / GMP A + B / D FED209 class 100
- Beskrivelse:CE bucket system trolley, without accessories, suitable for autoclaving and ISO Class 5
- Seq:12