VWR® Q Path® Coverquick, Mounting Agents for Microscopy
Q Path® Coverquick 1000, 260 mPaS, Plastic bottle, 500 mL
Leverandør: VWR Chemicals
VWR® Q Path® Coverquick, Mounting Agents for Microscopy
Katalog # 05547528.
Leverandør: VWR Chemicals
CAS nummer:
Gå til:
Product Details & Documents
A range of mounting media for manual and automatic mounting of slides. The Q Path® Coverquick range consists of four different mounting media with different viscosities to meet all microscopy needs. The number after the name indicates increasing viscosity: Q Path® Coverquick 1000 has the lower viscosity (around 260 mPaS) and Q Path® Coverquick 4000 has the higher viscosity (around 920 mPaS).
- A complete range of mounting media for histology and cytology
- The entire range is suitable for manual mounting
- For automatic mounting, Q Path® Coverquick 3000 and 4000 are recommended
- Delivered in PET amber bottle, only Q Path® Coverquick 3000 is delivered in a aluminium bottle
- Dries quickly in 15 to 20 min
Specifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 mL
- Beskrivelse:Q Path® Coverquick 1000, 260 mPaS