VWR® Q Path®, Medium-acting decalcifiers
DC2 medium-acting decalcifier, 2,5 L
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
This medium-acting decalcifier contains a mixture of hydrochloric acids.
- No formalin in the mixture
- Decalcify larger tissues already fixated
- Recommended volume is 20× the fragment volume
Ready to use for applications in histology and pathology laboratories.
Documents
Certifikater
Søg efter partinummerP20_VWRQ11028304_SPEC_EN.pdf spec_VWRQ11028304 spec_VWRQ11028304 spec_VWRQ11028304 spec_VWRQ11028306
Produktspecifikation
More
Specifikationer
Specifikationer
- Pakn:2,5 L
- Beskrivelse:DC2 medium-acting decalcifier