Orders LinkContactUsLinkComponent
VWR® Q Path®, Medium-acting decalcifiers
VWR® Q Path®, Medium-acting decalcifiers
DC2 medium-acting decalcifier, 2,5 L
Katalog # 11028306.
Leverandør:   VWR Chemicals
VWR® Q Path®, Medium-acting decalcifiers
Katalog # 11028306.
Leverandør:  VWR Chemicals
CAS nummer:  
VWR® Q Path®, Medium-acting decalcifiers
VWR® Q Path®, Medium-acting decalcifiers
DC2 medium-acting decalcifier, 2,5 L
CAS nummer:  
Price
792,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
This medium-acting decalcifier contains a mixture of hydrochloric acids.
  • No formalin in the mixture
  • Decalcify larger tissues already fixated
  • Recommended volume is 20× the fragment volume
Ready to use for applications in histology and pathology laboratories.
Specifikationer
Specifikationer
  • Pakn:
    2,5 L
  • Beskrivelse:
    DC2 medium-acting decalcifier