Accessories
Clutch plate, All Miele glassware units
Katalog # 141-0838
Gå til:
Discontinued
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifikationer
- Beskrivelse:Clutch plate
- Seq:07
- Til:All Miele glassware units