Inserts
E 430/1 insert, 1/3 mesh tray, for various instruments, wire gauge with mesh size 5 mm, for upper and lower basket, G 7881, G 7891, 150×445×40 mm
Katalog # 141-0525
Leverandør: MIELE
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:E 430/1 insert, 1/3 mesh tray, for various instruments, wire gauge with mesh size 5 mm, for upper and lower basket
- Seq:14
- B×D×H:150×445×40 mm
- Til:G 7881, G 7891