Accessories for comparators
pH, NLC, 6,0 - 7,6 pH
Katalog # LOVI281030
Leverandør: Lovibond
Gå til:
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specifikationer
- Område:6,0 - 7,6 pH
- Seq:57
- Kode:NLC
- Testmetode:pH