Absorbent pads and rolls, Universal Plus
87 L, 100 pads/box, 400×500 mm
Katalog # BRDY813842
Leverandør: Brady
Gå til:
Product Details & Documents
These absorbent pads and rolls are used when chemicals and hazardous liquids are involved.
- 2-ply meltblown and spunbound on one side, dimpled and perforated
- Absorb a wide range of chemicals, including acids and caustics
- One-sided spunbond gives extra strength as well as lower lint
- Perforated for flexibility – tear at the perforations for smaller sized pads
Chemically inert product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid. Green colour alerts workers that product is being used for hazardous liquids. Green colour also allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs.
Specifikationer
- Kapacitet:87 L
- L×B:400×500 mm
- Seq:01
- Pakket:100 pads/box
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



