Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pipette tips, ART™

Pipette tips, ART™

ART™ 10, 10 µl, -, 31,3 mm, 10 racks, 96 each
Katalog # 613-2464
   Lav fremstillingspåvirkning
Leverandør:   Thermo Fisher Scientific
Pipette tips, ART™
Katalog # 613-2464
   Lav fremstillingspåvirkning
Pipette tips, ART™

ART™ 10, 10 µl, -, 31,3 mm, 10 racks, 96 each

Price
368,00 kr
Udsalg

Gælder indtil kl 30/06/2026

Log ind for tilgængelighed
1 * 960 Items
Product Details & Documents

These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.

  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks ensure safe and secure storage
  • Space saving reload system is environmentally friendly, traceable and easy to use

Packaging: Tips are available in bulk packaging, in racks or in trays for rack reloading. Reload tips require an empty rack.

Specifikationer
  • Pakket:
    10 racks, 96 each
  • Steril:
    -
  • Volumen:
    10 µl
  • Længde:
    31,3 mm
  • Seq:
    02
  • Spidstype:
    ART™ 10
  • Cat. No.:
    613-2464
  • Miljøcertificering:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning
Accessories

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Tomt rack til refillpipettespidser, Medium
    Tomt rack til refillpipettespidser, Medium
    732-1170
    Log ind for tilgængelighed
    1 * 10 Items - 273,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Tomt rack til refillpipettespidser, Large
    Tomt rack til refillpipettespidser, Large
    732-1169
    Log ind for tilgængelighed
    1 * 8 Items - 134,00 kr

Produktinformation

  • Katalog #
    Tilgængelighed
    Price
  • Tomt rack til refillpipettespidser, Small
    Tomt rack til refillpipettespidser, Small
    732-1171
    Log ind for tilgængelighed
    1 * 10 Items - 153,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.