Orders LinkContactUsLinkComponent
Inoculating loops, Rotilabo®
Inoculating loops, Rotilabo®
Inoculating loops, platinum/iridium wire Ø 0,6 mm (without holder), 3 mm
Katalog # ROTHKN02.1
Marketsource
Leverandør:   Roth Carl
Inoculating loops, Rotilabo®
Katalog # ROTHKN02.1
Leverandør:  Roth Carl
Marketsource
Inoculating loops, Rotilabo®
Inoculating loops, platinum/iridium wire Ø 0,6 mm (without holder), 3 mm
Price
874,46 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Rotilabo® inoculating loops, made of platinum/iridium. Wire diameter 0,6 mm. Rotilabo® inoculating loop holder set made of stainless steel. Loops are fixed with union nut. For inoculating loops with wire diameter 0,6 to 1,0 mm. Set sorted by colour: blue, yellow, red, black. Length: 245 mm. Autoclavable at 121 °C.
Leveringsinformation: Loops delivered without inoculating loop holder, please order separately.
Specifikationer
  • Ø:
    3 mm
  • Beskrivelse:
    Inoculating loops, platinum/iridium wire Ø 0,6 mm (without holder)
  • Seq:
    02