Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, straight bore, 1,5 mm
Katalog # BIBBSST102K
Leverandør: SciLabware
Gå til:
Product Details & Documents
Fully interchangeable PTFE keys for Pyrex® stopcocks.
- Supplied complete with plastic screw thread retaining device
- Self lubricating, avoiding the need for stopcock grease
- Made to a taper of 1:10 giving a combination of performance and ease of operation
Delivery information: Key is supplied complete with plastic screw thread retaining device.
Specifikationer
- Hul Ø:1,5 mm
- Beskrivelse:Spare PTFE keys for stopcocks, PYREX®, straight bore
- Seq:01