Orders LinkContactUsLinkComponent
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, straight bore, 1,5 mm
Katalog # BIBBSST102K
Leverandør:   SciLabware
Spare PTFE Keys for Stopcocks, PYREX®
Katalog # BIBBSST102K
Leverandør:  SciLabware
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, straight bore, 1,5 mm
Price
875,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Fully interchangeable PTFE keys for Pyrex® stopcocks.
  • Supplied complete with plastic screw thread retaining device
  • Self lubricating, avoiding the need for stopcock grease
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
Delivery information: Key is supplied complete with plastic screw thread retaining device.
Specifikationer
  • Hul Ø:
    1,5 mm
  • Beskrivelse:
    Spare PTFE keys for stopcocks, PYREX®, straight bore
  • Seq:
    01