Orders LinkContactUsLinkComponent
Level control systems with signalboxes, SymLine
Level control systems with signalboxes, SymLine
Signalbox T5, 5-channel, universal plug, 180×105×55 mm
  SCTV108088
 :   S.C.A.T.
Level control systems with signalboxes, SymLine
  SCTV108088
 :  S.C.A.T.
Level control systems with signalboxes, SymLine
Level control systems with signalboxes, SymLine
Signalbox T5, 5-channel, universal plug, 180×105×55 mm
5.310,00 kr
1 * 1 Items
Monitor two filling levels at once, perfect integration into lab furniture with the Built-In Signalbox ant the Signalbox T1 and T4.
  • For all types of Safety Waste Caps and other electronic level controllers
  • Touchless alarm-resetting fields for two containers
  • 2 switch inputs and 2 switch outputs for peripheral units
LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via the integrated interface.
  • Beskrivelse:
    Signalbox T5, 5-channel, universal plug
  • Seq:
    02
  • B×D×H:
    180×105×55 mm
Accessories
  •  
    Beskrivelse
  • SCTV108304
    Signal cable cable for signal transmission, 1,5 m
    1 * 1 Items - 277,00 kr
  •  
    Beskrivelse
  • 590-0740
    EU power supply
    1 * 1 Items - 593,00 kr
  •  
    Beskrivelse
  • 554-2714
    Dual lock (releasable pressure) closure for disc sensors, 20×20 mm
    1 * 10 Items - 265,00 kr
  •  
    Beskrivelse
  • SCTV610703
    UK power supply
    1 * 1 Items - 593,00 kr
  •  
    Beskrivelse
  • 554-2715
    Velcro strip for disc sensor, 2 m
    1 * 1 Items - 239,00 kr
  •  
    Beskrivelse
  • 590-0727
    Forlængerkabel til signaloverførsel, 5 m
    1 * 1 Items - 425,00 kr
  •  
    Beskrivelse
  • 590-0728
    Forlængerkabel til signaloverførsel, 10 m
    1 * 1 Items - 443,00 kr
  •  
    Beskrivelse
  • 590-0729
    Kabel til relækontakt, 5 m
    1 * 1 Items - 357,00 kr
  •  
    Beskrivelse
  • 590-0734
    Disc sensor (alarm at empty state)
    1 * 1 Items - 2.300,00 kr
  •  
    Beskrivelse
  • 590-0737
    Niveausensor, enkelt (alarm i fuld tilstand)
    1 * 1 Items - 2.300,00 kr
  •  
    Beskrivelse
  • 590-0739
    Forlængerkabel til signaloverførsel, 3 m
    1 * 1 Items - 398,00 kr
  •  
    Beskrivelse
  • SCTV106731
    Holder for disc sensor for safety waste cap S60/61, three levels adjustable, PE_HD
    1 * 1 Items - 480,00 kr
  •  
    Beskrivelse
  • SCTV108334
    Signal lamp for level control, visual signal (flashing) when the level is reached, with magnetic holder for individual attachment, cable length of the lamp 3 m, power supply: 24 V DC EU/UK/US plug
    1 * 1 Items - 1.910,00 kr
  •  
    Beskrivelse
  • SCTV106723
    Holder for disc sensor, GL 45
    1 * 1 Items - 480,00 kr
  •  
    Beskrivelse
  • SCTV108334-T
    Signal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectors
    1 * 1 Items - 1.910,00 kr
  •  
    Beskrivelse
  • SCTV108335-T
    Signal lamp (full state) - EU/UK/US, for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment with disc sensor, visual alarm at full state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plug
    1 * 1 Items - 3.540,00 kr
  •  
    Beskrivelse
  • SCTV108336-T
    Signal lamp (empty state) - EU/UK/US, for level control, visual and acoustic signal at empty state, with magnetic holder for individual attachment with disc sensor, visual alarm at empty state, 3 m signal cable, 2 m hook and loop fastener for disc sensor; power supply: 24 V DC EU/UK/US plug
    1 * 1 Items - 3.540,00 kr