Pipette tips, ClickSure™ Tips
Standard, 50 µl, -, 10 racks, 96 each
Katalog # 613-2930
Leverandør: HAMILTON BONADUZ
Gå til:
Product Details & Documents
These tips are validated to optimise the performance of the Microlab 300 system, offering superior accuracy and precision. When the tips are placed onto the probe, the O-ring on the probe compresses into the tip groove. This design only requires a low insertion force to successfully load a tip, resulting in a more ergonomic experience for the user and enabling long-term use with minimal energy exerted.
- Tip packaging and tips are recyclable for eco-conscious labs
- Less force is required to load and eject a tip
Specifikationer
- Pakket:10 racks, 96 each
- Steril:-
- Volumen:50 µl
- Seq:02
- Spidstype:Standard