Silikoneslanger
Size 17, with hose barb of 6,3 mm, 6,40 mm, 17
SPECACTU-E17-25N
Silicone, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Rør int.Ø:6,40 mm
- Beskrivelse:Size 17, with hose barb of 6,3 mm
- Seq:03
- Størrelse:17