Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Threaded fittings

Container connection, PP, 25,4 mm, NW 24 mm, grey, PP, 25,4 mm
Katalog # BURK8530-0100
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8530-0100
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Container connection, PP, 25,4 mm, NW 24 mm, grey, PP, 25,4 mm

Price
153,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    25,4 mm
  • Beskrivelse:
    Container connection, PP, 25,4 mm, NW 24 mm, grey
  • Materiale:
    PP
  • Seq:
    30
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.