Metanil gul
Glass bottle for solids, 20 kg
Katalog # A17527.PG
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
CAS No.: 587-98-4 Molecular Formula: C18H14N3NaO3S Formula Weight: 375.38 Odour: Odourless pH: ~10 Beilstein Reference: 3831568 Merck Reference: 14,5928 MDL No.: MFCD00007486
Synonymer: Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
- CAS nummer:587-98-4
- Formel:C₁₈H₁₄N₃NaO₃S
- MDL nummer:MFCD00007486
- Molecular Weight:375.38 g/mol
- Merck Index:14,5928
- Opbevaringstemperatur:Stue temperatur
- EINECS:209-608-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:20 kg



