Visitor coats, PP
hvid, XXL
Katalog # 113-8330
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These disposable coats made of PP are ideal for visitors in non hazardous environments.
- With collar
- Press-stud fastening (5 press-studs)
- No pockets
Weight: 35 g/m²
Colour: White
Documents
Food Contact, BPA, Phthalates Certificate
Yderligere produktdetaljer
Specifikationer
- Tøj størrelse:XXL
- Farve:hvid
- Seq:06
- Køn:Unisex
- Cat. No.:113-8330
- Vægt:35 g/m²
- Outfit applikationer:General Purpose|Laboratory|Food
- Lukkestørrelse:Med trykknapper
- Lommer:uden lommer



