Visitor coats, PP
hvid, XL
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable coats made of PP are ideal for visitors in non hazardous environments.
- With collar
- Press-stud fastening (5 press-studs)
- No pockets
Weight: 35 g/m²
Colour: White
Documents
Food Contact, BPA, Phthalates Certificate
Additional Product Details
Specifications
- Tøj størrelse:XL
- Farve:hvid
- Seq:05
- Køn:Unisex
- Cat. No.:113-8329
- Vægt:35 g/m²
- Outfit applikationer:General Purpose|Laboratory|Food
- Lukkestørrelse:Med trykknapper
- Lommer:uden lommer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



