Visitor coats, PP
hvid, M
113-9261
These disposable coats made of PP are ideal for visitors in non hazardous environments.
- With collar
- Press-stud fastening (5 press-studs)
- No pockets
Weight: 35 g/m²
Colour: White
- Tøj størrelse:M
- Farve:hvid
- Seq:03
- Køn:Unisex
- Cat. No.:113-9261
- Vægt:35 g/m²
- Outfit applikationer:General Purpose|Laboratory|Food
- Lukkestørrelse:Med trykknapper
- Lommer:uden lommer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



