Shoe covers, CPE
Shoecover, CPE 40, blå, 150 mm, 410 mm
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Shoe covers made from CPE.
Documents
Certifikater
Søg efter partinummerFood Contact, BPA, Phthalates Certificate
Additional Product Details
Specifikationer
- Farve:blå
- Beskrivelse:Shoecover, CPE 40
- Højde:150 mm
- Længde:410 mm
- Seq:02