Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Shoe covers, CPE

Shoe covers, CPE

Shoecover, CPE 25, blå, 150 mm, 410 mm
Katalog # 113-9337
Leverandør:   VWR Collection
Shoe covers, CPE
Katalog # 113-9337
Leverandør:  VWR Collection
Shoe covers, CPE
Shoe covers, CPE

Shoecover, CPE 25, blå, 150 mm, 410 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 2.000 Items
Specifikationer
  • Farve:
    blå
  • Beskrivelse:
    Shoecover, CPE 25
  • Højde:
    150 mm
  • Længde:
    410 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.