Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Shoe covers, CPE

Shoe covers, CPE

Shoecover, CPE 70, grøn, 150 mm, 410 mm
Catalog # 113-9343
Supplier:   VWR Collection
Shoe covers, CPE
Catalog # 113-9343
Supplier:  VWR Collection
Shoe covers, CPE
Shoe covers, CPE

Shoecover, CPE 70, grøn, 150 mm, 410 mm

Price
Inquire For Price
Log in for availability
1 * 1.000 Items
Specifications
  • Farve:
    grøn
  • Beskrivelse:
    Shoecover, CPE 70
  • Højde:
    150 mm
  • Længde:
    410 mm
  • Seq:
    06