Orders LinkContactUsLinkComponent
Cross pieces, type KF
Cross pieces, type KF
Reducing 4-way cross, DN 25/16 KF, Rustfrit stål
Katalog # ILMV701601
Leverandør:   Welch by Gardner Denver
Cross pieces, type KF
Katalog # ILMV701601
Cross pieces, type KF
Reducing 4-way cross, DN 25/16 KF, Rustfrit stål
Price
975,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
It provides a flange fitting type.
Specifikationer
  • Beskrivelse:
    Reducing 4-way cross, DN 25/16 KF
  • Materiale:
    Rustfrit stål