Orders LinkContactUsLinkComponent
Centring rings, type KF
Centring rings, type KF
Centring rings with viton-O-ring, stainless steel, type KF, DN 16
Katalog # 181-0545
Leverandør:   Welch by Gardner Denver
Centring rings, type KF
Katalog # 181-0545
Centring rings, type KF
Centring rings with viton-O-ring, stainless steel, type KF, DN 16
Price
65,70 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Centring rings with viton-O-ring, stainless steel, type KF
  • Seq:
    04
  • Størrelse:
    DN 16