Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for AgilePulse™ MAX™

Accessories for AgilePulse™ MAX™

Large volume chamber, 6 mm gap, 10 ml
Katalog # BTXI47-0206
Leverandør:   BTX
Accessories for AgilePulse™ MAX™
Katalog # BTXI47-0206
Leverandør:  BTX
Accessories for AgilePulse™ MAX™
Accessories for AgilePulse™ MAX™

Large volume chamber, 6 mm gap, 10 ml

Price
14.800,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Large volume chamber, 6 mm gap, 10 ml
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.