Orders LinkContactUsLinkComponent
Mirrors, Magoptic
Mirrors, Magoptic
Mirror, with focal length of -200 mm, 40 mm
Katalog # DIDAPOD608717
Leverandør:   DIDALAB
Mirrors, Magoptic
Katalog # DIDAPOD608717
Leverandør:  DIDALAB
Mirrors, Magoptic
Mirrors, Magoptic
Mirror, with focal length of -200 mm, 40 mm
Price
58,80 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Mirrors used for enabling classroom experiments.
Specifikationer
  • Beskrivelse:
    Mirror, with focal length of -200 mm
  • Seq:
    03
  • Ø:
    40 mm