Orders LinkContactUsLinkComponent
Mirrors, Magoptic
Mirrors, Magoptic
Mirror, with focal length of +100 mm, 80 mm
Katalog # DIDAPOD608414
Leverandør:   DIDALAB
Mirrors, Magoptic
Katalog # DIDAPOD608414
Leverandør:  DIDALAB
Mirrors, Magoptic
Mirrors, Magoptic
Mirror, with focal length of +100 mm, 80 mm
Price
111,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Mirrors used for enabling classroom experiments.
Specifikationer
  • Beskrivelse:
    Mirror, with focal length of +100 mm
  • Seq:
    10
  • Ø:
    80 mm