Orders LinkContactUsLinkComponent
Screens for optical bench
Screens for optical bench
Translucent screen 20×20 cm, attached to a rod with Ø 10 mm, one graduated side
Katalog # DIDAPOD010007
Leverandør:   DIDALAB
Screens for optical bench
Katalog # DIDAPOD010007
Leverandør:  DIDALAB
Screens for optical bench
Screens for optical bench
Translucent screen 20×20 cm, attached to a rod with Ø 10 mm, one graduated side
Price
216,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Translucent screen 20×20 cm, attached to a rod with Ø 10 mm, one graduated side