Screens for optical bench
Translucent screen 20×20 cm, attached to a rod with Ø 10 mm, one graduated side
Katalog # DIDAPOD010007
Leverandør: DIDALAB
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Translucent screen 20×20 cm, attached to a rod with Ø 10 mm, one graduated side