Microplates, 384-well, non binding
384-well microplate, non binding, flat μClear® bottom, hvid/klar
Katalog # 738-0176
Leverandør: Greiner Bio-One
Microplates, 384-well, non binding
Katalog # 738-0176
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
All plates are free from RNase, DNase, and pyrogens, and are also free of DNA polymerase inhibitors.
Specifikationer
- Farve:hvid/klar
- Beskrivelse:384-well microplate, non binding, flat μClear® bottom
- Seq:05
- Cat. No.:738-0176
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lav fremstillingspåvirkning