Orders LinkContactUsLinkComponent
Mop heads, DryMop™
Mop heads, DryMop™
Dry Mop™ handle, metal, 1524 mm
Katalog # MICRDMH-60M
Leverandør:   MICRONOVA
Mop heads, DryMop™
Katalog # MICRDMH-60M
Leverandør:  MICRONOVA
Mop heads, DryMop™
Mop heads, DryMop™
Dry Mop™ handle, metal, 1524 mm
Price
505,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
For dusting and dry mopping large surface areas.
  • Low profile design
  • Polyester pad head limits particle generation
  • Swivel head for easy maneuvering
  • Launderable/Autoclavable covers
Specifikationer
  • Beskrivelse:
    Dry Mop™ handle, metal
  • Længde:
    1524 mm
  • Seq:
    04