Orders LinkContactUsLinkComponent
RNase A
RNase A
400 µl
Katalog # AC117
Leverandør:   OMEGA BIO-TEK
RNase A
Katalog # AC117
Leverandør:  OMEGA BIO-TEK
CAS nummer:  
RNase A
400 µl
CAS nummer:  
Price
255,00 kr
Log ind for tilgængelighed
1 * 400 µl
Product Details & Documents
Ribonuclease A (RNase A) is endonuclease-free and quality controlled for use in plasmid purification procedures for digestion of RNA.
Specifikationer
Specifikationer
  • Pakn:
    400 µl
  • Cat. No.:
    AC117
  • Koncentration:
    25 mg/ml
  • Enzym navn:
    RNase A
Specifikationer