Orders LinkContactUsLinkComponent
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40, with standard blade
Katalog # SWAN2580
Leverandør:   SWANN MORTON
Disposable Post Mortem scalpels
Katalog # SWAN2580
Leverandør:  SWANN MORTON
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40, with standard blade
Price
917,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Carbon blade. Non sterile, disposable histopathology and autopsy knives.
  • With easy to remove guard to protect the cutting edge: Users do not have to fix the blade to the handle
  • Available with either standard PM40 blade or PM40B bull nose blade fixed in a distinctive blue handle
Documents
Specifikationer
  • Model:
    PM40, with standard blade