Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Affinity purification resins, anti-His, GenScript®

Affinity purification resins, anti-His, GenScript®

Anti-His affinity resin, 50% resin in TBS with 0,02% sodium azide, Liquid, 5 mL
Katalog # GENSL00439-5
Leverandør:   Genscript
Affinity purification resins, anti-His, GenScript®
Katalog # GENSL00439-5
Leverandør:  Genscript
CAS nummer:  
Affinity purification resins, anti-His, GenScript®

Anti-His affinity resin, 50% resin in TBS with 0,02% sodium azide, Liquid, 5 mL

CAS nummer:  
Price
20.220,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents

Anti-His affinity resins are designed for purification of His-tagged protein. Composed of agarose and conjugated THE™ anti-His monoclonal antibody, they are compatible with most commonly used reagents at certain concentrations.

  • Beads structure: 4% agarose
  • Beads size: 45 to 165 μm, average 90 μm
  • Binding capacity: greater than 3 mg of human C-reactive protein per ml of settled resin
  • Specificity: N-terminal/C-terminal/internal/4 x His/5 x His/6 x His-tagged protein
  • Storage: 2 to 8 °C (avoid freezing the resin)
  • Binding condition: Tris or borate-buffered saline (pH 8 to 8,5) with 1 to 2 mM calcium chloride
  • Elution methods: Acid, alkaline and peptide competitive elutions

His-tagged protein in the crude samples can bind to the anti-His monoclonal antibodies coupled with the agarose, and the non specific proteins can be eliminated by washing. As a result, the His-tagged protein will finally be eluted with high recovery. Moreover, this product can be used for immunoprecipitation of His-tagged protein.

Generally the resin can be recycled at least four times. If maintained properly, the resin may be recycled 10 times with minor binding capability loss.

Specifikationer

Specifikationer

  • Pakn:
    5 mL
  • Beskrivelse:
    Anti-His affinity resin, 50% resin in TBS with 0,02% sodium azide
  • Model:
    Liquid

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.