Hand towels, SCOTT® PERFORMANCE
SCOTT® Performance, 1 ply, medium, hvid, 315×215 mm, 15×212 towels
Katalog # KIMW6663
Leverandør: KIMBERLY CLARK
Product Details & Documents
These 1-ply interfolded hand towels are made from strong, highly absorbent AIRFLEX* fabric and supplied in cost-saving ‘compressed’ packs (with 18% more towels per case).
- Soft yet highly absorbent, even when wet
- Dry hands hygienically and efficiently
Ideal for busy office and hotel washrooms.
Specifikationer
- Pakket:15×212 towels
- Farve:hvid
- Beskrivelse:SCOTT® Performance, 1 ply, medium
- L×B:315×215 mm
- Seq:03
Complementary Products
Produktinformation
- Katalog #BeskrivelseB×D×HTilgængelighedPrice
- 115-1780Dispenser for C-fold hand towels, AQUARIUS* 6945, white ABS, with window, large280×146×412 mmLog ind for tilgængelighed1 * 1 Items - 415,00 kr
- 115-1783Dispenser for interfolded hand towels, KIMBERLY-CLARK PROFESSIONAL*, 8971, stainless steel237×118×349 mmLog ind for tilgængelighed1 * 1 Items - 2.850,00 kr
- 115-1782Dispenser for C-fold hand towels, AQUARIUS* 6956, white ABS, with window, small307×176×155 mmLog ind for tilgængelighed1 * 1 Items - 277,00 kr