Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pipette controller, Pipet-Aid® XP2

Pipette controller, Pipet-Aid® XP2

Pipet-Aid® XP2, CE, with International plug, 240 V, 1 - 100 ml
Katalog # DRUM4-000-501-I
Leverandør:   DRUMMOND
Pipette controller, Pipet-Aid® XP2
Katalog # DRUM4-000-501-I
Leverandør:  DRUMMOND
Pipette controller, Pipet-Aid® XP2

Pipet-Aid® XP2, CE, with International plug, 240 V, 1 - 100 ml

Price
9.570,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

This controller is an advanced version of the Pipet-Aid® XP controller and accepts 1 to 100 ml pipettes.

  • New ergonomic design
  • New power source for uninterrupted extended operation; can be charged while in use
  • Ultra quiet precision pump provides great control for aspiration or dispensing

Leveringsinformation: The unit is supplied complete with a power supply/charger, four extra filters, and a removable stand.

Specifikationer
  • Kapacitet:
    1 - 100 ml
  • Beskrivelse:
    Pipet-Aid® XP2, CE, with International plug, 240 V
  • Seq:
    01