Orders LinkContactUsLinkComponent
VWR® Q Path® Coverquick, Mounting Agents for Microscopy
VWR® Q Path® Coverquick, Mounting Agents for Microscopy
Q Path® Coverquick 3000, 610 mPaS, Aluminium bottle, 500 mL
Katalog # 05547537.
Leverandør:   VWR Chemicals
VWR® Q Path® Coverquick, Mounting Agents for Microscopy
Katalog # 05547537.
Leverandør:  VWR Chemicals
CAS nummer:  
VWR® Q Path® Coverquick, Mounting Agents for Microscopy
VWR® Q Path® Coverquick, Mounting Agents for Microscopy
Q Path® Coverquick 3000, 610 mPaS, Aluminium bottle, 500 mL
CAS nummer:  
Price
648,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
A range of mounting media for manual and automatic mounting of slides. The Q Path® Coverquick range consists of four different mounting media with different viscosities to meet all microscopy needs. The number after the name indicates increasing viscosity: Q Path® Coverquick 1000 has the lower viscosity (around 260 mPaS) and Q Path® Coverquick 4000 has the higher viscosity (around 920 mPaS).
  • A complete range of mounting media for histology and cytology
  • The entire range is suitable for manual mounting
  • For automatic mounting, Q Path® Coverquick 3000 and 4000 are recommended
  • Delivered in PET amber bottle, only Q Path® Coverquick 3000 is delivered in a aluminium bottle
  • Dries quickly in 15 to 20 min
Specifikationer
Specifikationer
  • Pakning:
    Aluminium bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    Q Path® Coverquick 3000, 610 mPaS