VWR®, Forceps
Sterile, 120 mm, Individually wrapped
Leverandør: VWR Collection
Gå til:
Product Details & Documents
PS, translucent blue.
- Combines high mechanical strength, flexibility and chemical resistance
- Available in sterile (individually wrapped) and non sterile versions
Documents
CoC VWR Forceps, PS, translucent blue plastic with pointed end.pdf CoC VWR Forceps, PS, Translucent Blue Plastic with Pointed Ends.pdf CoC VWR Forceps.pdf CoC VWR, Forceps.pdf CoC_VWR_FORCEPS_BLUE_TRANSLUCENT.pdf
Overensstemmelseserklæring
COC_VWRI232-0191_20180910 CoC_VWRI232-0191_20190320 COC_VWRI232-0191_20190610 CoC_VWRI232-0191_20190820 COC_VWRI232-0191_20190920
More
TDS_VWR_FORCEPS_BLUE_TRANSLUCENT.pdf
Produktspecifikation
VWRI232-0191_TDS_EN
Datablad
VWR® Lab bric-a-brac
Produktlitteratur
Specifikationer
- Pakket:Individually wrapped
- Model:Sterile
- Længde:120 mm
- Seq:01
- Cat. No.:232-0191
- Miljøcertificering:ISO 14001 (Certificate Registration No.131120)
- Environmentally Preferable:Lav fremstillingspåvirkning