Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human recombinant SDCBP

Human recombinant SDCBP

25 µG
Katalog # ABNOH00006386-P012
Leverandør:   Abnova
Human recombinant SDCBP
Katalog # ABNOH00006386-P012
Leverandør:  Abnova
CAS nummer:  
Human recombinant SDCBP

25 µG

CAS nummer:  
Price
4.040,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human SDCBP full-length ORF recombinant protein with GST-tag at N-terminal.

  • Host: Wheat germ (in vitro)
  • Purifification: by Glutathione sepharose 4 fast flow
  • Storage buffer: 50 mM Tris-HCI, 10 mM reduced Glutathione, pH = 8,0 in the elution buffer
  • Preparation method: in vitro wheat germ expression system
  • Quality control testing: 12,5% SDS-PAGE stained with coomassie blue

Used in Enzyme-linked immunoabsorbent assay, western blot (recombinant protein), antibody production and protein array.

Specifikationer

Specifikationer

  • Pakn:
    25 µG
  • Cat. No.:
    ABNOH00006386-P012
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    SDCBP
  • Molekylvægt:
    58,8 kDA
  • Species:
    Human
  • Opbevaring betingelser:
    –80 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.