Dropping bottles, glass
20 ml, DIN 18
Katalog # HECH41314020
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Made of amber glass.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Amber coloured to protect light sensitive substances
Dropping bottles without cap, with thread DIN 18.
: Pipette kit and screw cap must be ordered seperately.
Specifikationer
- Gevind:DIN 18
- Kapacitet:20 ml
- Seq:02
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 5 ml dropping bottlesPipette kit, red, for 5 ml dropping bottlesHECH41315006Log ind for tilgængelighed1 * 100 Items - kr662.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, blue, for 20 ml dropping bottlesPipette kit, blue, for 20 ml dropping bottlesHECH41315020Log ind for tilgængelighed1 * 100 Items - kr643.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, blue, for 30 ml dropping bottlesPipette kit, blue, for 30 ml dropping bottlesHECH41315030Log ind for tilgængelighed1 * 100 Items - kr662.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 30 ml dropping bottlesPipette kit, red, for 30 ml dropping bottlesHECH41315031Log ind for tilgængelighed1 * 100 Items - kr662.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 100 ml dropping bottlesPipette kit, red, for 100 ml dropping bottlesHECH41315061Log ind for tilgængelighed1 * 100 Items - kr674.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 20 ml dropping bottlesPipette kit, red, for 20 ml dropping bottlesHECH41315021Log ind for tilgængelighed1 * 100 Items - kr643.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 10 ml dropping bottlesPipette kit, red, for 10 ml dropping bottlesHECH41315011Log ind for tilgængelighed1 * 100 Items - kr643.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, red, for 50 ml dropping bottlesPipette kit, red, for 50 ml dropping bottlesHECH41315051Log ind for tilgængelighed1 * 100 Items - kr653.00
Produktinformation
- Katalog #TilgængelighedPrice
- Pipette kit, blue, for 50 ml dropping bottlesPipette kit, blue, for 50 ml dropping bottlesHECH41315650Log ind for tilgængelighed1 * 100 Items - kr466.00