Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Drenchers

Drenchers

Drencher, bent, 102 mm, 1,6 mm
Katalog # SOCO376.16102B
Leverandør:   Socorex Isba
Drenchers
Katalog # SOCO376.16102B
Leverandør:  Socorex Isba
Drenchers
Drenchers

Drencher, bent, 102 mm, 1,6 mm

Price
187,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents

These straight or bent drenchers are made from stainless steel. They are ideal for safe animal nose and mouth feeding.

  • Luer lock fittings
Specifikationer
  • Udv.Ø:
    1,6 mm
  • Beskrivelse:
    Drencher, bent
  • Længde:
    102 mm
  • Seq:
    03
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.