Orders LinkContactUsLinkComponent
Control Cables
Control Cables
Control cable to connect units with RS232 (15 pol.) to PC, 9 pol. sub-D, 3 m
Katalog # HUBE55018
Leverandør:   HUBER
Control Cables
Katalog # HUBE55018
Leverandør:  HUBER
Control Cables
Control Cables
Control cable to connect units with RS232 (15 pol.) to PC, 9 pol. sub-D, 3 m
Price
1.300,00 kr
Log ind for tilgængelighed
1 * 3 m
Product Details & Documents
A range of control cables is available for USB, RS232, RS485 or analogue interfaces (AIF). Cables are also available for external control signal (ECS), a potentialfree contact (POKO) or for an external float switch (LEVEL).
Specifikationer
  • Beskrivelse:
    Control cable to connect units with RS232 (15 pol.) to PC, 9 pol. sub-D
  • Længde:
    3 m
  • Seq:
    02