Gowning bench
Gowning bench, L×W: 1524×406 mm, 28,4 kg
Katalog # METOGB1660S
Leverandør: Metro International
Gå til:
Product Details & Documents
Gowning bench in stainless steel 304.
- Heavy-duty, solid top bench provides comfortable gowning fixture
- Stainless steel foot plates provide floor mount or free-standing option
Documents
METOGB1636S_TDS_EN
Datablad
Specifikationer
- Beskrivelse:Gowning bench, L×W: 1524×406 mm
- Seq:03
- Vægt:28,4 kg