Gowning bench
Gowning bench, L×W: 914×406 mm, 21,5 kg
Catalog # METOGB1636S
Supplier: Metro International
Jump to:
Product Details & Documents
Gowning bench in stainless steel 304.
- Heavy-duty, solid top bench provides comfortable gowning fixture
- Stainless steel foot plates provide floor mount or free-standing option
Documents
METOGB1636S_TDS_EN
Datasheet
Specifications
- Beskrivelse:Gowning bench, L×W: 914×406 mm
- Seq:01
- Vægt:21,5 kg