Cleanroom labels, matte
Removable cleanroom labels, matte finish, PP, Akryl, hvid, 76×51 mm, 1000 labels/roll
Katalog # 817-0305
Leverandør: Texwipe
Gå til:
Product Details & Documents
Matte finish cleanroom labels provide excellent compatibility with cleanroom pens and thermal transfer inks. Available with two different adhesive levels to provide excellent adhesion to a variety of surfaces without leaving residue.
- Ideal for handwritten applications
Specifikationer
- Farve:hvid
- Beskrivelse:Removable cleanroom labels, matte finish
- Dimensioner:76×51 mm
- Materiale:PP
- Seq:02
- Pakket:1000 labels/roll
- Klæbestof:Akryl