Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
TRItidy G™

TRItidy G™

TRItidy G™, 100 mL
Katalog # A4051.0100
Leverandør:   PanReac AppliChem
TRItidy G™
Katalog # A4051.0100
Leverandør:  PanReac AppliChem
CAS nummer:  
TRItidy G™
TRItidy G™

TRItidy G™, 100 mL

CAS nummer:  
Price
1.260,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

TRItidy G™ is based on the single step simultaneous isolation of RNA, DNA and proteins from cell and tissue samples (Chomczynski, P. (1993), BioTechniques 15, p.532 - 537). RNA is selectively retained in the aqueous phase during acidic GuSCN/phenol extraction; DNA and proteins are isolated from the organic phase by ethanol and isopropanol precipitation, respectively.

  • Ready to use
  • Suitable for samples (tissues and cells) of human, animal, plant and bacterial origin
  • Isolation of large and small RNA species (0,1 - 15 kb) with high purity

Documents

Specifikationer

Specifikationer

  • Pakn:
    100 mL
  • Beskrivelse:
    TRItidy G™
  • Cat. No.:
    A4051.0100
  • Prep size:
    Scalable
  • Beskrivelse:
    TRItidy G™
  • Refine target molecule:
    Total RNA|Total DNA|Total protein
  • Target molecule:
    DNA|RNA|Protein|Co-purification of RNA, DNA and protein
  • Method / Format:
    Precipitation|Organic extraction
  • Sample type:
    Cultured cells|Invertebrates|Human/animal cells and tissues

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.