Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
TRItidy G™

TRItidy G™

TRItidy G™, 200 mL
Katalog # A4051.0200
Leverandør:   PanReac AppliChem
TRItidy G™
Katalog # A4051.0200
Leverandør:  PanReac AppliChem
CAS nummer:  
TRItidy G™
TRItidy G™

TRItidy G™, 200 mL

CAS nummer:  
Price
2.550,00 kr
Log ind for tilgængelighed
1 * 200 mL
Product Details & Documents

TRItidy G™ is based on the single step simultaneous isolation of RNA, DNA and proteins from cell and tissue samples (Chomczynski, P. (1993), BioTechniques 15, p.532 - 537). RNA is selectively retained in the aqueous phase during acidic GuSCN/phenol extraction; DNA and proteins are isolated from the organic phase by ethanol and isopropanol precipitation, respectively.

  • Ready to use
  • Suitable for samples (tissues and cells) of human, animal, plant and bacterial origin
  • Isolation of large and small RNA species (0,1 - 15 kb) with high purity

Documents

Specifikationer

Specifikationer

  • Pakn:
    200 mL
  • Beskrivelse:
    TRItidy G™
  • Cat. No.:
    A4051.0200
  • Prep size:
    Scalable
  • Beskrivelse:
    TRItidy G™
  • Refine target molecule:
    Total RNA|Total DNA|Total protein
  • Target molecule:
    DNA|RNA|Protein|Co-purification of RNA, DNA and protein
  • Method / Format:
    Precipitation|Organic extraction
  • Sample type:
    Cultured cells|Invertebrates|Human/animal cells and tissues

Specifikationer