Orders LinkContactUsLinkComponent
Centrifugerør
50 ml, 34×100 mm, uden cap, Round bottom
Catalog # MARI3933041
Supplier:   MARIENFELD
Centrifugerør
Catalog # MARI3933041
Supplier:  MARIENFELD
Centrifugerør
Centrifugerør
50 ml, 34×100 mm, uden cap, Round bottom
Price
605,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Borosilicate glass 3,3.
  • Heavy walled
  • Ungraduated
  • Very good chemical resistance
  • Very good temperature resistance
Short conical or round bottom.
Specifications
  • Lukkestørrelse:
    uden cap
  • Ø×L:
    34×100 mm
  • Model:
    Round bottom
  • Kapacitet:
    50 ml
  • Seq:
    03